Assay Details
| Gene Symbol: PCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169632.2 | NP_001163103.1 | 5 - 6 | 2006 | 137 | |
| NM_165709.2 | NP_724841.1 | 4 - 5 | 1781 | 137 | ||
| NM_165710.3 | NP_724842.1 | 4 - 5 | 2078 | 137 | ||
| NM_165711.2 | NP_724843.1 | 4 - 5 | 1823 | 137 | ||
| NM_165712.2 | NP_724844.1 | 3 - 4 | 1888 | 137 | ||
More Information
Set Membership: |
|

PCB
Genomic Map