Assay Details
| Gene Symbol: Pka-R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103781.3 | NP_001097251.1 | 3 - 4 | 568 | 60 | |
| NM_001103782.2 | NP_001097252.1 | 3 - 4 | 725 | 60 | ||
| NM_078947.4 | NP_523671.1 | 3 - 4 | 568 | 60 | ||
| NM_165723.4 | NP_724860.2 | 3 - 4 | 589 | 60 | ||
| GenBank mRNA | AF274008.1 | - | 3 - 4 | 439 | 60 | |
| AY069669.1 | - | 3 - 4 | 439 | 60 | ||
| BT029610.1 | - | 3 - 4 | 701 | 60 | ||
| BT029637.1 | - | 3 - 4 | 701 | 60 | ||
| BT029963.1 | - | 2 - 3 | 372 | 60 | ||
More Information
Set Membership: |
|

Pka-R2
Genomic Map