Assay Details
| Gene Symbol: Pka-R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103781.3 | NP_001097251.1 | 5 - 6 | 846 | 96 | |
| NM_001103782.2 | NP_001097252.1 | 5 - 6 | 1003 | 96 | ||
| NM_078947.4 | NP_523671.1 | 5 - 6 | 846 | 96 | ||
| NM_165723.4 | NP_724860.2 | 5 - 6 | 867 | 96 | ||
| GenBank mRNA | AF274008.1 | - | 5 - 6 | 717 | 96 | |
| AY069669.1 | - | 5 - 6 | 717 | 96 | ||
| BT029610.1 | - | 5 - 6 | 979 | 96 | ||
| BT029637.1 | - | 5 - 6 | 979 | 96 | ||
| BT029963.1 | - | 4 - 5 | 650 | 96 | ||
More Information
Set Membership: |
|

Pka-R2
Genomic Map