Assay Details
| Gene Symbol: Pka-R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103781.3 | NP_001097251.1 | 7 - 8 | 1088 | 60 | |
| NM_001103782.2 | NP_001097252.1 | 7 - 8 | 1245 | 60 | ||
| NM_078947.4 | NP_523671.1 | 7 - 8 | 1088 | 60 | ||
| NM_165723.4 | NP_724860.2 | 7 - 8 | 1109 | 60 | ||
| GenBank mRNA | AF274008.1 | - | 7 - 8 | 959 | 60 | |
| AY069669.1 | - | 7 - 8 | 959 | 60 | ||
| BT029610.1 | - | 7 - 8 | 1221 | 60 | ||
| BT029637.1 | - | 7 - 8 | 1221 | 60 | ||
| BT029963.1 | - | 6 - 7 | 892 | 60 | ||
More Information
Set Membership: |
|

Pka-R2
Genomic Map