Assay Details
| Gene Symbol: Pka-R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103781.3 | NP_001097251.1 | 8 - 9 | 1170 | 84 | |
| NM_001103782.2 | NP_001097252.1 | 8 - 9 | 1327 | 84 | ||
| NM_078947.4 | NP_523671.1 | 8 - 9 | 1170 | 84 | ||
| NM_165723.4 | NP_724860.2 | 8 - 9 | 1191 | 84 | ||
| GenBank mRNA | AF274008.1 | - | 8 - 9 | 1041 | 84 | |
| AY069669.1 | - | 8 - 9 | 1041 | 84 | ||
| BT029610.1 | - | 8 - 9 | 1303 | 84 | ||
| BT029637.1 | - | 8 - 9 | 1303 | 84 | ||
| BT029963.1 | - | 7 - 8 | 974 | 84 | ||
More Information
Set Membership: |
|

Pka-R2
Genomic Map