Assay Details
| Gene Symbol: TER94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103779.2 | NP_001097249.1 | 4 - 5 | 2482 | 58 | |
| NM_001103780.2 | NP_001097250.1 | 4 - 5 | 2549 | 58 | ||
| NM_001299332.1 | NP_001286261.1 | 4 - 5 | 2479 | 58 | ||
| NM_058021.4 | NP_477369.1 | 4 - 5 | 2440 | 58 | ||
| GenBank mRNA | AF132553.1 | - | 4 - 5 | 2422 | 58 | |
More Information
Set Membership: |
|

TER94
Genomic Map