Assay Details
| Gene Symbol: Cdc2rk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078950.4 | NP_523674.1 | 3 - 4 | 649 | 84 | |
| GenBank mRNA | BT089018.1 | - | 3 - 4 | 606 | 84 | |
More Information
Set Membership: |
|

Cdc2rk
Genomic Map