Assay Details
| Gene Symbol: RanBPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038849.2 | NP_001033938.1 | 5 - 6 | 1989 | 93 | |
| NM_001259326.1 | NP_001246255.1 | 5 - 6 | 1992 | 93 | ||
| NM_001273938.1 | NP_001260867.1 | 5 - 6 | 1640 | 93 | ||
| NM_001316413.1 | NP_001303342.1 | 5 - 6 | 1993 | 93 | ||
| NM_136741.3 | NP_610585.2 | 5 - 6 | 1643 | 93 | ||
| NM_165770.3 | NP_724932.1 | 5 - 6 | 1993 | 93 | ||
| GenBank mRNA | AY973545.1 | - | 5 - 6 | 1950 | 93 | |
| AY973546.1 | - | 5 - 6 | 1950 | 93 | ||
| BT150441.1 | - | 5 - 6 | 1643 | 93 | ||
More Information
Set Membership: |
|

RanBPM
Genomic Map