Assay Details
| Gene Symbol: RanBPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259326.1 | NP_001246255.1 | 9 - 10 | 2627 | 71 | |
| NM_001316413.1 | NP_001303342.1 | 9 - 10 | 2628 | 71 | ||
| NM_136741.3 | NP_610585.2 | 9 - 10 | 2278 | 71 | ||
| NM_165770.3 | NP_724932.1 | 9 - 10 | 2628 | 71 | ||
| GenBank mRNA | BT150441.1 | - | 9 - 10 | 2278 | 71 | |
More Information
Set Membership: |
|

RanBPM
Genomic Map