Assay Details
| Gene Symbol: dgo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136770.2 | NP_610614.1 | 3 - 4 | 682 | 79 | |
| NM_165800.2 | NP_724973.1 | 3 - 4 | 1079 | 79 | ||
| GenBank mRNA | AY128436.1 | - | 3 - 4 | 645 | 79 | |
More Information
Set Membership: |
|

dgo
Genomic Map