Assay Details
| Gene Symbol: Prosbeta5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143757.4 | NP_652014.1 | 1 - 2 | 241 | 77 | |
| NM_165801.2 | NP_724974.1 | 2 - 3 | 178 | 77 | ||
| GenBank mRNA | AY069381.1 | - | 1 - 2 | 219 | 77 | |
More Information
Set Membership: |
|

Prosbeta5
Genomic Map