Assay Details
| Gene Symbol: dhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297953.1 | NP_001284882.1 | 1 | 897 | 71 | |
| NM_078491.3 | NP_511046.1 | 1 | 714 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dhd| Gene Symbol: dhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297953.1 | NP_001284882.1 | 1 | 897 | 71 | |
| NM_078491.3 | NP_511046.1 | 1 | 714 | 71 | ||
Set Membership: |
|