Assay Details
| Gene Symbol: swa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078505.4 | NP_511060.2 | 2 - 3 | 1228 | 64 | |
| GenBank mRNA | AY069487.1 | - | 2 - 3 | 1229 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

swa| Gene Symbol: swa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078505.4 | NP_511060.2 | 2 - 3 | 1228 | 64 | |
| GenBank mRNA | AY069487.1 | - | 2 - 3 | 1229 | 64 | |
Set Membership: |
|