Assay Details
| Gene Symbol: PpV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272343.1 | NP_001259272.1 | 2 - 3 | 483 | 69 | |
| NM_001272344.1 | NP_001259273.1 | 2 - 3 | 479 | 69 | ||
| NM_001272345.1 | NP_001259274.1 | 3 - 4 | 502 | 69 | ||
| NM_078506.3 | NP_511061.1 | 2 - 3 | 555 | 69 | ||
| GenBank mRNA | BT021230.1 | - | 2 - 3 | 545 | 69 | |
| X75980.1 | - | 481 | 69 | |||
More Information
Set Membership: |
|

PpV
Genomic Map