Assay Details
| Gene Symbol: CBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272380.2 | NP_001259309.1 | 1 - 2 | 504 | 62 | |
| NM_078515.4 | NP_511070.1 | 1 - 2 | 504 | 62 | ||
| GenBank mRNA | AF104405.1 | - | 1 - 2 | 483 | 62 | |
| AF145675.1 | - | 1 - 2 | 486 | 62 | ||
More Information
Set Membership: |
|

CBP
Genomic Map