Assay Details
| Gene Symbol: sws | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078520.3 | NP_511075.3 | 3 - 4 | 2463 | 76 | |
| NM_167140.2 | NP_727224.2 | 3 - 4 | 2463 | 76 | ||
| GenBank mRNA | BT133162.1 | - | 3 - 4 | 2463 | 76 | |
More Information
Set Membership: |
|

sws
Genomic Map