Assay Details
| Gene Symbol: sn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169226.2 | NP_001162697.1 | 2 - 3 | 556 | 71 | |
| NM_078521.3 | NP_511076.3 | 2 - 3 | 427 | 71 | ||
| NM_167142.3 | NP_727226.1 | 3 - 4 | 690 | 71 | ||
| NM_167143.3 | NP_727227.1 | 2 - 3 | 947 | 71 | ||
More Information
Set Membership: |
|

sn
Genomic Map