Assay Details
| Gene Symbol: fs(1)h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272392.2 | NP_001259321.1 | 9 - 10 | 6542 | 78 | |
| NM_078523.3 | NP_511078.2 | 9 - 10 | 6518 | 78 | ||
More Information
Set Membership: |
|

fs(1)h
Genomic Map