Assay Details
| Gene Symbol: org-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298090.1 | NP_001285019.1 | 4 - 5 | 1712 | 80 | |
| NM_078530.3 | NP_511085.2 | 4 - 5 | 1512 | 80 | ||
More Information
Set Membership: |
|

org-1
Genomic Map