Assay Details
| Gene Symbol: rdgA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042799.2 | NP_001036264.2 | 11 - 12 | 2923 | 64 | |
| NM_001103446.2 | NP_001096916.1 | 10 - 11 | 2566 | 64 | ||
| NM_001103447.4 | NP_001096917.1 | 12 - 13 | 4169 | 64 | ||
| NM_001201635.1 | NP_001188564.1 | 11 - 12 | 2923 | 64 | ||
| NM_001201636.1 | NP_001188565.1 | 11 - 12 | 4778 | 64 | ||
| NM_001272437.1 | NP_001259366.1 | 11 - 12 | 4808 | 64 | ||
| NM_001272438.1 | NP_001259367.1 | 11 - 12 | 4802 | 64 | ||
| NM_001298098.1 | NP_001285027.1 | 11 - 12 | 4808 | 64 | ||
| NM_001298099.1 | NP_001285028.1 | 11 - 12 | 4808 | 64 | ||
| NM_001382089.1 | NP_001368977.1 | 11 - 12 | 4808 | 64 | ||
| NM_078537.3 | NP_511092.2 | 11 - 12 | 4793 | 64 | ||
| GenBank mRNA | AY075349.1 | - | 11 - 12 | 4796 | 64 | |
More Information
Set Membership: |
|

rdgA
Genomic Map