Assay Details
| Gene Symbol: rdgA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042799.2 | NP_001036264.2 | 13 - 14 | 3198 | 66 | |
| NM_001103447.4 | NP_001096917.1 | 14 - 15 | 4444 | 66 | ||
| NM_001201636.1 | NP_001188565.1 | 13 - 14 | 5053 | 66 | ||
| NM_001272437.1 | NP_001259366.1 | 13 - 14 | 5083 | 66 | ||
| NM_001272438.1 | NP_001259367.1 | 13 - 14 | 5077 | 66 | ||
| NM_078537.3 | NP_511092.2 | 13 - 14 | 5068 | 66 | ||
| GenBank mRNA | AY075349.1 | - | 13 - 14 | 5071 | 66 | |
More Information
Set Membership: |
|

rdgA
Genomic Map