Assay Details
| Gene Symbol: rdgA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042799.2 | NP_001036264.2 | 14 - 15 | 3262 | 60 | |
| NM_001103446.2 | NP_001096916.1 | 14 - 15 | 2959 | 60 | ||
| NM_001103447.4 | NP_001096917.1 | 15 - 16 | 4508 | 60 | ||
| NM_001201635.1 | NP_001188564.1 | 15 - 16 | 3316 | 60 | ||
| NM_001201636.1 | NP_001188565.1 | 14 - 15 | 5117 | 60 | ||
| NM_001272437.1 | NP_001259366.1 | 14 - 15 | 5147 | 60 | ||
| NM_001272438.1 | NP_001259367.1 | 14 - 15 | 5141 | 60 | ||
| NM_001298099.1 | NP_001285028.1 | 13 - 14 | 5095 | 60 | ||
| NM_078537.3 | NP_511092.2 | 14 - 15 | 5132 | 60 | ||
| GenBank mRNA | AY075349.1 | - | 14 - 15 | 5135 | 60 | |
More Information
Set Membership: |
|

rdgA
Genomic Map