Assay Details
| Gene Symbol: rdgA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042799.2 | NP_001036264.2 | 8 - 9 | 1985 | 74 | |
| NM_001103446.2 | NP_001096916.1 | 7 - 8 | 1628 | 74 | ||
| NM_001103447.4 | NP_001096917.1 | 9 - 10 | 3231 | 74 | ||
| NM_001201635.1 | NP_001188564.1 | 8 - 9 | 1985 | 74 | ||
| NM_001201636.1 | NP_001188565.1 | 8 - 9 | 3840 | 74 | ||
| NM_001272438.1 | NP_001259367.1 | 8 - 9 | 3864 | 74 | ||
| NM_078537.3 | NP_511092.2 | 8 - 9 | 3855 | 74 | ||
| GenBank mRNA | AY075349.1 | - | 8 - 9 | 3858 | 74 | |
More Information
Set Membership: |
|

rdgA
Genomic Map