Assay Details
| Gene Symbol: Ost48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078541.4 | NP_511096.2 | 1 - 2 | 231 | 73 | |
| GenBank mRNA | BT010050.1 | - | 1 - 2 | 208 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ost48| Gene Symbol: Ost48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078541.4 | NP_511096.2 | 1 - 2 | 231 | 73 | |
| GenBank mRNA | BT010050.1 | - | 1 - 2 | 208 | 73 | |
Set Membership: |
|