Assay Details
| Gene Symbol: lz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103449.2 | NP_001096919.1 | 2 - 3 | 1085 | 64 | |
| NM_078544.3 | NP_511099.2 | 2 - 3 | 1085 | 64 | ||
| GenBank mRNA | DQ397338.1 | - | 1 - 2 | 863 | 64 | |
More Information
Set Membership: |
|

lz
Genomic Map