Assay Details
| Gene Symbol: Yp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298142.1 | NP_001285071.1 | 1 - 2 | 324 | 86 | |
| NM_078548.3 | NP_511103.1 | 1 - 2 | 283 | 86 | ||
More Information
Set Membership: |
|

Yp1
Genomic Map