Assay Details
| Gene Symbol: Imp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078556.4 | NP_511111.1 | 3 - 4 | 1000 | 89 | |
| NM_167249.3 | NP_727451.1 | 3 - 4 | 1260 | 89 | ||
| NM_167250.3 | NP_727452.1 | 4 - 5 | 1411 | 89 | ||
| NM_167251.3 | NP_727453.1 | 3 - 4 | 1621 | 89 | ||
| NM_167252.3 | NP_727454.1 | 3 - 4 | 1725 | 89 | ||
| NM_167253.3 | NP_727455.1 | 4 - 5 | 1617 | 89 | ||
| NM_167254.4 | NP_727456.1 | 3 - 4 | 779 | 89 | ||
| NM_167255.3 | NP_727457.1 | 3 - 4 | 973 | 89 | ||
| GenBank mRNA | AF241237.1 | - | 3 - 4 | 888 | 89 | |
| AY069821.1 | - | 3 - 4 | 742 | 89 | ||
More Information
Set Membership: |
|

Imp
Genomic Map