Assay Details
| Gene Symbol: Imp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042803.3 | NP_001036268.2 | 5 - 6 | 1875 | 66 | |
| NM_001169246.2 | NP_001162717.1 | 4 - 5 | 2349 | 66 | ||
| NM_001258687.3 | NP_001245616.1 | 5 - 6 | 2627 | 66 | ||
| NM_001298162.1 | NP_001285091.1 | 5 - 6 | 2231 | 66 | ||
| NM_078556.4 | NP_511111.1 | 4 - 5 | 1707 | 66 | ||
| NM_167249.3 | NP_727451.1 | 4 - 5 | 1967 | 66 | ||
| NM_167250.3 | NP_727452.1 | 5 - 6 | 2118 | 66 | ||
| NM_167251.3 | NP_727453.1 | 4 - 5 | 2328 | 66 | ||
| NM_167252.3 | NP_727454.1 | 4 - 5 | 2432 | 66 | ||
| NM_167253.3 | NP_727455.1 | 5 - 6 | 2324 | 66 | ||
| NM_167254.4 | NP_727456.1 | 4 - 5 | 1486 | 66 | ||
| NM_167255.3 | NP_727457.1 | 4 - 5 | 1680 | 66 | ||
| GenBank mRNA | AF241237.1 | - | 4 - 5 | 1595 | 66 | |
| BT133404.1 | - | 4 - 5 | 2346 | 66 | ||
More Information
Set Membership: |
|

Imp
Genomic Map