Assay Details
| Gene Symbol: Imp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042803.3 | NP_001036268.2 | 6 - 7 | 2028 | 81 | |
| NM_001169246.2 | NP_001162717.1 | 5 - 6 | 2502 | 81 | ||
| NM_001258687.3 | NP_001245616.1 | 6 - 7 | 2780 | 81 | ||
| NM_001298162.1 | NP_001285091.1 | 6 - 7 | 2384 | 81 | ||
| NM_078556.4 | NP_511111.1 | 5 - 6 | 1860 | 81 | ||
| NM_167249.3 | NP_727451.1 | 5 - 6 | 2120 | 81 | ||
| NM_167250.3 | NP_727452.1 | 6 - 7 | 2271 | 81 | ||
| NM_167251.3 | NP_727453.1 | 5 - 6 | 2481 | 81 | ||
| NM_167252.3 | NP_727454.1 | 5 - 6 | 2585 | 81 | ||
| NM_167253.3 | NP_727455.1 | 6 - 7 | 2477 | 81 | ||
| NM_167254.4 | NP_727456.1 | 5 - 6 | 1639 | 81 | ||
| NM_167255.3 | NP_727457.1 | 5 - 6 | 1833 | 81 | ||
| GenBank mRNA | AF241237.1 | - | 5 - 6 | 1748 | 81 | |
| AY069821.1 | - | 5 - 6 | 1602 | 81 | ||
| BT133404.1 | - | 5 - 6 | 2499 | 81 | ||
More Information
Set Membership: |
|

Imp
Genomic Map