Assay Details
| Gene Symbol: sev | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078559.3 | NP_511114.2 | 4 - 5 | 1053 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sev| Gene Symbol: sev | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078559.3 | NP_511114.2 | 4 - 5 | 1053 | 107 | |
Set Membership: |
|