Assay Details
| Gene Symbol: hop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078564.3 | NP_511119.2 | 5 - 6 | 2847 | 105 | |
| GenBank mRNA | BT006331.1 | - | 5 - 6 | 2847 | 105 | |
More Information
Set Membership: |
|

hop
Genomic Map