Assay Details
| Gene Symbol: hop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078564.3 | NP_511119.2 | 8 - 9 | 3815 | 98 | |
| GenBank mRNA | BT006331.1 | - | 8 - 9 | 3815 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hop| Gene Symbol: hop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078564.3 | NP_511119.2 | 8 - 9 | 3815 | 98 | |
| GenBank mRNA | BT006331.1 | - | 8 - 9 | 3815 | 98 | |
Set Membership: |
|