Assay Details
| Gene Symbol: RpII215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078569.3 | NP_511124.1 | 1 - 2 | 444 | 86 | |
| GenBank mRNA | BT100316.1 | - | 1 - 2 | 440 | 86 | |
More Information
Set Membership: |
|

RpII215
Genomic Map