Assay Details
| Gene Symbol: nod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298200.1 | NP_001285129.1 | 2 - 3 | 469 | 71 | |
| NM_078570.3 | NP_511125.2 | 2 - 3 | 469 | 71 | ||
| GenBank mRNA | AY050238.1 | - | 2 - 3 | 423 | 71 | |
More Information
Set Membership: |
|

nod
Genomic Map