Assay Details
| Gene Symbol: gd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272549.1 | NP_001259478.1 | 3 - 4 | 604 | 63 | |
| NM_001272550.1 | NP_001259479.1 | 3 - 4 | 613 | 63 | ||
| NM_001316623.1 | NP_001303552.1 | 3 - 4 | 604 | 63 | ||
| GenBank mRNA | AF056311.1 | - | 3 - 4 | 588 | 63 | |
More Information
Set Membership: |
|

gd
Genomic Map