Assay Details
| Gene Symbol: mew | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272575.2 | NP_001259504.1 | 1 - 2 | 563 | 87 | |
| NM_001298267.1 | NP_001285196.1 | 1 - 2 | 563 | 87 | ||
| NM_078590.3 | NP_511145.2 | 1 - 2 | 563 | 87 | ||
More Information
Set Membership: |
|

mew
Genomic Map