Assay Details
| Gene Symbol: mew | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272574.1 | NP_001259503.1 | 3 - 4 | 307 | 70 | |
| NM_001298267.1 | NP_001285196.1 | 2 - 3 | 698 | 70 | ||
| NM_078590.3 | NP_511145.2 | 2 - 3 | 698 | 70 | ||
| NM_167354.2 | NP_727679.1 | 3 - 4 | 307 | 70 | ||
| GenBank mRNA | AY075338.1 | - | 3 - 4 | 307 | 70 | |
More Information
Set Membership: |
|

mew
Genomic Map