Assay Details
| Gene Symbol: Yp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298299.1 | NP_001285228.1 | 1 - 2 | 265 | 57 | |
| NM_078593.4 | NP_511148.2 | 1 - 2 | 265 | 57 | ||
More Information
Set Membership: |
|

Yp3
Genomic Map