Assay Details
| Gene Symbol: mRpS25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078598.2 | NP_511153.1 | 2 - 3 | 421 | 88 | |
| GenBank mRNA | BT024442.1 | - | 2 - 3 | 366 | 88 | |
| BT029396.1 | - | 2 - 3 | 421 | 88 | ||
More Information
Set Membership: |
|

mRpS25
Genomic Map