Assay Details
| Gene Symbol: rut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258743.3 | NP_001245672.1 | 16 - 17 | 6055 | 76 | |
| NM_001272619.2 | NP_001259548.1 | 16 - 17 | 6328 | 76 | ||
| NM_001272620.2 | NP_001259549.1 | 15 - 16 | 6341 | 76 | ||
| NM_001272621.2 | NP_001259550.1 | 15 - 16 | 4922 | 76 | ||
| NM_001272622.2 | NP_001259551.1 | 15 - 16 | 5270 | 76 | ||
| NM_001298317.1 | NP_001285246.1 | 16 - 17 | 6403 | 76 | ||
| NM_078601.4 | NP_511156.2 | 17 - 18 | 6634 | 76 | ||
More Information
Set Membership: |
|

rut
Genomic Map