Assay Details
| Gene Symbol: rut | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258743.3 | NP_001245672.1 | 5 - 6 | 1399 | 77 | |
| NM_001272619.2 | NP_001259548.1 | 5 - 6 | 1399 | 77 | ||
| NM_001272620.2 | NP_001259549.1 | 4 - 5 | 1337 | 77 | ||
| NM_001272621.2 | NP_001259550.1 | 5 - 6 | 1447 | 77 | ||
| NM_001272622.2 | NP_001259551.1 | 5 - 6 | 1447 | 77 | ||
| NM_001298317.1 | NP_001285246.1 | 5 - 6 | 1399 | 77 | ||
| NM_078601.4 | NP_511156.2 | 5 - 6 | 1399 | 77 | ||
More Information
Set Membership: |
|

rut
Genomic Map