Assay Details
| Gene Symbol: dah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078607.3 | NP_511162.1 | 2 - 3 | 552 | 87 | |
| GenBank mRNA | BT016120.1 | - | 2 - 3 | 546 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dah| Gene Symbol: dah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078607.3 | NP_511162.1 | 2 - 3 | 552 | 87 | |
| GenBank mRNA | BT016120.1 | - | 2 - 3 | 546 | 87 | |
Set Membership: |
|