Assay Details
| Gene Symbol: dah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272639.2 | NP_001259568.1 | 4 - 5 | 1513 | 80 | |
| NM_078607.3 | NP_511162.1 | 4 - 5 | 1522 | 80 | ||
| GenBank mRNA | BT016120.1 | - | 4 - 5 | 1516 | 80 | |
More Information
Set Membership: |
|

dah
Genomic Map