Assay Details
| Gene Symbol: dah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272639.2 | NP_001259568.1 | 5 - 6 | 1674 | 62 | |
| NM_078607.3 | NP_511162.1 | 5 - 6 | 1683 | 62 | ||
| GenBank mRNA | BT016120.1 | - | 5 - 6 | 1677 | 62 | |
More Information
Set Membership: |
|

dah
Genomic Map