Assay Details
| Gene Symbol: exd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272663.2 | NP_001259592.1 | 2 - 3 | 528 | 74 | |
| NM_001272664.1 | NP_001259593.1 | 2 - 3 | 481 | 74 | ||
| NM_078636.5 | NP_523360.1 | 2 - 3 | 497 | 74 | ||
| NM_167478.3 | NP_727923.1 | 2 - 3 | 458 | 74 | ||
| GenBank mRNA | AY061065.1 | - | 2 - 3 | 526 | 74 | |
| L19295.1 | - | 2 - 3 | 472 | 74 | ||
More Information
Set Membership: |
|

exd
Genomic Map