Assay Details
| Gene Symbol: exd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272663.2 | NP_001259592.1 | 3 - 4 | 846 | 67 | |
| NM_001272664.1 | NP_001259593.1 | 3 - 4 | 799 | 67 | ||
| NM_078636.5 | NP_523360.1 | 3 - 4 | 815 | 67 | ||
| NM_167478.3 | NP_727923.1 | 3 - 4 | 776 | 67 | ||
| GenBank mRNA | AY061065.1 | - | 3 - 4 | 844 | 67 | |
| L19295.1 | - | 3 - 4 | 790 | 67 | ||
More Information
Set Membership: |
|

exd
Genomic Map