Assay Details
| Gene Symbol: mei-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078645.3 | NP_523369.2 | 2 - 3 | 2501 | 63 | |
| GenBank mRNA | AY282465.1 | - | 2 - 3 | 2501 | 63 | |
More Information
Set Membership: |
|

mei-41
Genomic Map