Assay Details
| Gene Symbol: AnxB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272685.1 | NP_001259614.1 | 6 - 7 | 1032 | 56 | |
| NM_001272686.1 | NP_001259615.1 | 5 - 6 | 963 | 56 | ||
| NM_001272687.1 | NP_001259616.1 | 7 - 8 | 1543 | 56 | ||
| NM_001272688.1 | NP_001259617.1 | 6 - 7 | 978 | 56 | ||
| NM_001272689.1 | NP_001259618.1 | 7 - 8 | 1985 | 56 | ||
| NM_078646.3 | NP_523370.1 | 6 - 7 | 1032 | 56 | ||
| NM_167519.2 | NP_727978.1 | 6 - 7 | 1530 | 56 | ||
| GenBank mRNA | AJ276963.1 | - | 6 - 7 | 1006 | 56 | |
| AY058397.1 | - | 6 - 7 | 1006 | 56 | ||
| AY122138.2 | - | 6 - 7 | 980 | 56 | ||
| BT125828.1 | - | 6 - 7 | 1531 | 56 | ||
More Information
Set Membership: |
|

AnxB11
Genomic Map