Assay Details
| Gene Symbol: mbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078651.4 | NP_523375.2 | 1 - 2 | 1676 | 64 | |
| GenBank mRNA | AJ011578.1 | - | 1 - 2 | 1625 | 64 | |
| BT044204.1 | - | 1 - 2 | 1649 | 64 | ||
More Information
Set Membership: |
|

mbt
Genomic Map