Assay Details
| Gene Symbol: RpS19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298409.1 | NP_001285338.1 | 2 - 3 | 105 | 51 | |
| NM_001298410.1 | NP_001285339.1 | 1 - 2 | 163 | 51 | ||
| NM_078652.4 | NP_523376.1 | 2 - 3 | 105 | 51 | ||
| NM_167526.3 | NP_727992.1 | 2 - 3 | 127 | 51 | ||
| NM_167527.3 | NP_727993.1 | 1 - 2 | 410 | 51 | ||
| GenBank mRNA | AY118868.1 | - | 2 - 3 | 120 | 51 | |
More Information
Set Membership: |
|

RpS19a
Genomic Map